Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylmaleimide, 99%, Thermo Scientific™

Catalog No. AAA1355122
Change view
Click to view available options
Quantity:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1355122 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1355122 Supplier Thermo Scientific Chemicals Supplier No. A1355122

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Benzylmaleimide is used as plastic additive, Pharmaceutical Intermediates. It is an important raw material intermediate used in organic Synthesis, Pharmaceuticals, Agrochemicals Dyestuff.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1631-26-1
Molecular Formula C11H9NO2
Quantity 100g
MDL Number MFCD00014540
UN Number UN1759
Beilstein 135801
Synonym n-benzylmaleimide, 1-benzyl-1h-pyrrole-2,5-dione, n-benzyl maleimide, 1h-pyrrole-2,5-dione, 1-phenylmethyl, 1-benzylmaleimide, n-benzylmaleinimide, 1-benzyl-pyrrole-2,5-dione, n-benzyl-maleimide, 1-benzyl-2,5-dihydro-1h-pyrrole-2,5-dione, n-benzylmaleimide dihydrate
Solubility Information Insoluble in water.
InChI Key MKRBAPNEJMFMHU-UHFFFAOYSA-N
SMILES O=C1C=CC(=O)N1CC1=CC=CC=C1
IUPAC Name 1-benzyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Weight (g/mol) 187.20
PubChem CID 74204
Formula Weight 187.2
Percent Purity 99%
Chemical Name or Material N-Benzylmaleimide
Show More Show Less
Hazard Category H301-H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 216-631-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.