Learn More
Medchemexpress LLC N-Benzyloxycarbonyl-2-phosphonoglycine trimethyl ester | 88568-95-0 | 99.6% | 331.26 | 25 G
SDP
Catalog No. 5000403094
Encompass_Preferred
Trimethyl N-(benzyloxycarbonyl)-2-phosphonoglycinate is a biochemical reagent used as a biological material or organic compound for life science research. It is for research use only.
  • Used as a biological material or organic compound for life science related research.
  • Starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes.
  • Dehydroamino acids can be stereoselectively reduced to amino acids.
Catalog No. 50-004-03094 Supplier Medchemexpress LLC Supplier No. 25GHY33291
Trimethyl N-(benzyloxycarbonyl)-2-phosphonoglycinate is a biochemical reagent used as a biological material or organic compound for life science research. It is for research use only.
  • Used as a biological material or organic compound for life science related research.
  • Starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes.
  • Dehydroamino acids can be stereoselectively reduced to amino acids.