Learn More
Medchemexpress LLC N-(Benzyloxycarbonyl)-D-homoserine | 41088-85-1 | MFCD12545887 | 253.25 | C12H15NO5 | 1 G
SDP
Catalog No. 5000299642
Encompass_Preferred
N-(Benzyloxycarbonyl)-D-homoserine is the D-enantiomer of homoserine bearing an N-carbobenzoxy (Cbz) protecting group. Supplied as a powder, it is used as a drug intermediate in small-molecule synthesis and can be employed in the preparation of antibody-drug conjugates (ADCs). Molecular formula C12H15NO5; molecular weight 253.25. Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Protected amino group enables selective synthetic transformations.
  • Suitable for use as a drug intermediate in medicinal chemistry workflows.
  • Powder form supports convenient handling and long-term storage.
  • Defined molecular formula and mass aid analytical characterization.
  • Established storage recommendations help preserve compound stability.
Catalog No. 50-002-99642 Supplier Medchemexpress LLC Supplier No. HYW025723A1G
N-(Benzyloxycarbonyl)-D-homoserine is the D-enantiomer of homoserine bearing an N-carbobenzoxy (Cbz) protecting group. Supplied as a powder, it is used as a drug intermediate in small-molecule synthesis and can be employed in the preparation of antibody-drug conjugates (ADCs). Molecular formula C12H15NO5; molecular weight 253.25. Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Protected amino group enables selective synthetic transformations.
  • Suitable for use as a drug intermediate in medicinal chemistry workflows.
  • Powder form supports convenient handling and long-term storage.
  • Defined molecular formula and mass aid analytical characterization.
  • Established storage recommendations help preserve compound stability.