Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-D-serine, 98+%, Thermo Scientific™

Catalog No. AAL0838403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0838403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0838403 Supplier Thermo Scientific Chemicals Supplier No. L0838403

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6081-61-4
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.227
MDL Number MFCD00063144
InChI Key GNIDSOFZAKMQAO-SECBINFHSA-N
Synonym z-d-ser-oh, n-cbz-d-serine, cbz-d-ser-oh, z-d-serine, n-carbobenzoxy-d-serine, n-carbobenzyloxy-d-serine, r-2-benzyloxy carbonyl amino-3-hydroxypropanoic acid, cbz-d-serine, n-benzyloxycarbonyl-d-serine, benzyloxycarbonyl-d-serine
PubChem CID 489184
IUPAC Name (2R)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 116°C to 119°C
Quantity 1 g
Beilstein 2058313
Optical Rotation −6° (c=7 in Acetic Acid)
Formula Weight 239.23
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-D-serine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.