Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-DL-asparagine, 98%, Thermo Scientific™

Catalog No. AAH6658722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6658722 100 g
AAH6658714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6658722 Supplier Thermo Scientific Chemicals Supplier No. H6658722

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29880-22-6
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.253
MDL Number MFCD00065101
InChI Key FUCKRCGERFLLHP-UHFFFAOYSA-N
Synonym z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine
PubChem CID 246565
IUPAC Name 4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O

Specifications

Color White
Beilstein 6066596
Quantity 100 g
Formula Weight 266.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-DL-asparagine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.