Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-DL-valine, 98%, Thermo Scientific™

Catalog No. AAH6678314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6678314 25 g
AAH6678306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6678314 Supplier Thermo Scientific Chemicals Supplier No. H6678314

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3588-63-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00065127
InChI Key CANZBRDGRHNSGZ-UHFFFAOYSA-N
Synonym n-carbobenzoxy-dl-valine, z-dl-val-oh, n-cbz-dl-valine, 2-benzyloxy carbonyl amino-3-methylbutanoic acid, n-carbobenzoyl-dl-valine, n-benzyloxy carbonyl valine, n-benzyloxycarbonyl-dl-valine, n-carbobenzyl-oxy-dl-valine, dl-valine, n-phenylmethoxy carbonyl
PubChem CID 97743
IUPAC Name 2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
SMILES CC(C)C(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Beilstein 2056615
Quantity 25 g
Formula Weight 251.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-DL-valine
EINECSNumber 222-727-4
TSCA Yes
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.