Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH6364206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6364206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6364206 Supplier Thermo Scientific Chemicals Supplier No. H6364206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-47-2
Molecular Formula C13H15NO6
Molecular Weight (g/mol) 281.264
MDL Number MFCD00133585
InChI Key PHMBNDDHIBIDRQ-JTQLQIEISA-N
Synonym z-asp ome-oh, s-2-n-cbz-amino-succinic acid 4-methyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-methyl ester, s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, 2s-2-benzyloxy carbonyl amino-4-methoxy-4-oxobutanoic acid, cbz-l-aspartic acid 4-methyl ester, n-cbz-l-aspartate, pubchem14062, z-l-aspartic acid 4-methyl ester, z-asp ome-oh tlc
PubChem CID 7014888
IUPAC Name (2S)-4-methoxy-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES COC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Color White
Beilstein 6279422
Quantity 5 g
Formula Weight 281.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.