Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH5983806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5983806 5 g
AAH5983803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5983806 Supplier Thermo Scientific Chemicals Supplier No. H5983806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5545-52-8 | Molecular Formula: C16H21NO6 | Molecular Weight: 323.345 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in DMSO, and methanol.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5545-52-8
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00042857
InChI Key HLSLRFBLVZUVIE-LBPRGKRZSA-N
Synonym z-asp otbu-oh, n-cbz-l-aspartic acid 4-tert-butyl ester, cbz-l-aspartic acid 4-tert-butyl ester, cbz-asp obut-oh, z-asp otbu-oh.dcha, cbz-l-aspartic 4-tert-butyl ester, s-2-benzyloxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, z-l-aspartic acid 4-tert-butyl ester, 4-tert-butyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, z-asp otbu-oh.h2o
PubChem CID 111082
IUPAC Name (2S)-4-[(2-methylpropan-2-yl)oxy]-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Linear Formula (CH3)3CO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 5 g
Beilstein 2569846
Solubility Information Soluble in DMSO,and methanol.
Formula Weight 323.34
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester
EINECSNumber 226-912-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.