Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyloxycarbonyl-L-isoleucine dicyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAH6673322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6673322 100 g
AAH6673314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6673322 Supplier Thermo Scientific Chemicals Supplier No. H6673322

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26699-00-3
Molecular Formula C26H42N2O4
Molecular Weight (g/mol) 446.63
MDL Number MFCD00043243
InChI Key QWMHUFMEYKIYPC-QUTFAALHNA-N
Synonym cbz-l-isoleucine dicyclohexylamine salt, z-ile-oh dicyclohexylammonium salt, cbz-ile-oh.dcha, n-benzyloxy carbonyl-l-isoleucine, compound with dicyclohexylamine 1:1, cbz-isoleucine.dcha, cbz-ile-oh??dcha, pubchem18961, z-l-isoleucine dicyclohexylammonium salt, cbz-l-isoleucine dicyclohexylammonium salt, n-alpha-benzyloxycarbonyl-l-isoleucine dicyclohexylamine
PubChem CID 43835055
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoic acid; N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1.CCC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 159°C to 163°C
Color White
Beilstein 6465170
Quantity 100 g
Formula Weight 446.63
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Benzyloxycarbonyl-L-isoleucine dicyclohexylammonium salt
EINECSNumber 247-910-6
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.