Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-proline, 98+%

Click to view available options
Quantity:
5g
50g
250g
1148-11-4
C13H15NO4
249.266
MFCD00003170
JXGVXCZADZNAMJ-NSHDSACASA-N
z-pro-oh,n-benzyloxycarbonyl-l-proline,n-carbobenzoxy-l-proline,n-cbz-l-proline,benzyloxycarbonyl-l-proline,carbobenzoxyproline,carbobenzyloxy-l-proline,carbobenzoxy-l-proline,benzyloxycarbonylproline,n-carbobenzyloxy-l-proline
101987
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
1148-11-4
C13H15NO4
249.266
MFCD00003170
JXGVXCZADZNAMJ-NSHDSACASA-N
z-pro-oh,n-benzyloxycarbonyl-l-proline,n-carbobenzoxy-l-proline,n-cbz-l-proline,benzyloxycarbonyl-l-proline,carbobenzoxyproline,carbobenzyloxy-l-proline,carbobenzoxy-l-proline,benzyloxycarbonylproline,n-carbobenzyloxy-l-proline
101987
(2S)-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)O
Applications
N-(Benzyloxycarbonyl)-L-proline is a potent in vitro and in vivo inhibitor of prolidase; a specific peptidase that cleaves dipeptides with a C-terminal prolyl and hydroxylprolyl residue. It is also used in the synthesis of N-(L-Prolyl)-β-alanine which is a derivative of the naturally occurring beta amino acid β-Alanine.

Solubility
Solubility in methanol, almost transparency.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 75°C to 77°C
Quantity 250g
Optical Rotation −60° (c=1 in Acetic Acid)
Beilstein 88579
Formula Weight 249.27
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-proline

RTECSNumber : UY0745000