Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(benzyloxycarbonyl)-L-serine benzyl ester | 21209-51-8 | MFCD00037824 | 97.0% | 329.35 g/mol | C18H19NO5 | 50 G
SDP

Catalog No. 5000210382
Encompass_Preferred

Z-Ser-obzl is a Z-protected serine derivative supplied as a biochemical reagent for laboratory research use. It is typically used as a protected amino acid intermediate in peptide synthesis and related organic chemistry applications.

  • Purity 97.0%.
  • Molecular weight 329.35 g/mol.
  • Chemical formula C18H19NO5.
  • CAS number 21209-51-8 for unambiguous identification.
  • Available in multiple package sizes, including 50 G.
  • Intended for research use only; follow standard laboratory safety practices.

Catalog No. 50-002-10382 Supplier Medchemexpress LLC Supplier No. HYW04400150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Ser-obzl is a Z-protected serine derivative supplied as a biochemical reagent for laboratory research use. It is typically used as a protected amino acid intermediate in peptide synthesis and related organic chemistry applications.

  • Purity 97.0%.
  • Molecular weight 329.35 g/mol.
  • Chemical formula C18H19NO5.
  • CAS number 21209-51-8 for unambiguous identification.
  • Available in multiple package sizes, including 50 G.
  • Intended for research use only; follow standard laboratory safety practices.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.