Learn More
Cayman Chemical N-Benzylpalmitamide, Purity: ≥98%, CAS: 74058-71-2, 100 mg
SDP
Catalog No. 502892705
Encompass_Preferred
A long-chain fatty acid amide isolated from the maca plant that moderately inhibits FAAH activity (44% inhibition at 500 µM); many members of this family demonstrate selective antiproliferative activity against diverse cancer cell lines
Catalog No. 50-289-2705 Supplier Cayman Chemical Supplier No. 9002235100 MG
A long-chain fatty acid amide isolated from the maca plant that moderately inhibits FAAH activity (44% inhibition at 500 µM); many members of this family demonstrate selective antiproliferative activity against diverse cancer cell lines