Learn More
Cayman Chemical N-BenzylpalmItamIde 50mg
SDP
Catalog No. 502893615
Encompass_Preferred
A long-chain fatty acid amide isolated from the maca plant that moderately inhibits FAAH activity (44% inhibition at 500 µM); many members of this family demonstrate selective antiproliferative activity against diverse cancer cell lines
Catalog No. 50-289-3615 Supplier Cayman Chemical Supplier No. 900223550 MG
A long-chain fatty acid amide isolated from the maca plant that moderately inhibits FAAH activity (44% inhibition at 500 µM); many members of this family demonstrate selective antiproliferative activity against diverse cancer cell lines