Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyltropinone, 97%, Thermo Scientific™

Catalog No. MO07738DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07738DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07738DA Supplier Thermo Scientific Chemicals Supplier No. MO07738DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28957-72-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.30
MDL Number MFCD00151640
InChI Key RSUHKGOVXMXCND-UHFFFAOYNA-N
Synonym 8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
PubChem CID 119846
IUPAC Name 8-benzyl-8-azabicyclo[3.2.1]octan-3-one
SMILES O=C1CC2CCC(C1)N2CC1=CC=CC=C1

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.