Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(Beta-Ketocaproyl)-L-Homoserine Lactone

Catalog No. K300710MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K300710MG 50 mg
K300750MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K300710MG Supplier Sigma Aldrich Supplier No. K300710MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(Beta-Ketocaproyl)-L-Homoserine Lactone

Specifications

CAS 143537-62-6
Molecular Formula C10H15NO4
Linear Formula C10H15NO4
MDL Number MFCD00171363
Quantity 50 mg
Synonym N-(3-Oxohexanoyl)-L-homoserine lactone
Molecular Weight (g/mol) 213.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.