Learn More
Echelon Biosciences Research Labs N-BIOTIN CERAMIDE 1-PHOSPHATE 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs S500B50UG
Encompass
Ceramide 1-phosphate (C1P) is biosynthesized from ceramide by the intracellular enzyme Ceramide kinase. C1P inhibits cell apoptosis via inhibition of acid sphingomyelinase activity in contrast to ceramide which promotes apoptosis. C1P is important in inflammation, activating cytosolic phospholipase A2 alpha and releasing downstream inflammatory mediators. In addition, C1P is involved in DNA synthesis, mast cell degranulation and neutrophil phagocytosis. This item is non returnable
Catalog No. 50-822-67