Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-biotinyl-1,6-hexanediamine | 65953-56-2 | 98.2% | 342.5 g/mol | C16H30N4O2S | 50 MG
SDP

Catalog No. 5000282204
Encompass_Preferred

N-biotinyl-1,6-hexanediamine is a biotinylated hexylamine reagent used to introduce a biotin tag via a six-carbon spacer for labeling, affinity capture, and conjugation in biochemical assays and synthetic chemistry. It is supplied as an off-white to pale yellow solid and is commonly employed as a synthetic intermediate or labeling reagent in life-science research.

  • Provides a biotin tag for affinity capture and detection.
  • Six-carbon spacer reduces steric hindrance for conjugation.
  • High purity (~98.2%) suitable for biochemical applications.
  • Molecular weight 342.5 g/mol and defined chemical formula C16H30N4O2S.
  • Soluble in DMSO and methanol; limited solubility in chloroform and ethanol.
  • Off-white to pale yellow solid with melting point 184-186 °C.
  • Store powder at -20 °C; store solutions at -80 °C when indicated.

Catalog No. 50-002-82204 Supplier Medchemexpress LLC Supplier No. HYW34359750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-biotinyl-1,6-hexanediamine is a biotinylated hexylamine reagent used to introduce a biotin tag via a six-carbon spacer for labeling, affinity capture, and conjugation in biochemical assays and synthetic chemistry. It is supplied as an off-white to pale yellow solid and is commonly employed as a synthetic intermediate or labeling reagent in life-science research.

  • Provides a biotin tag for affinity capture and detection.
  • Six-carbon spacer reduces steric hindrance for conjugation.
  • High purity (~98.2%) suitable for biochemical applications.
  • Molecular weight 342.5 g/mol and defined chemical formula C16H30N4O2S.
  • Soluble in DMSO and methanol; limited solubility in chloroform and ethanol.
  • Off-white to pale yellow solid with melting point 184-186 °C.
  • Store powder at -20 °C; store solutions at -80 °C when indicated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.