Learn More
Medchemexpress LLC N-biotinyl-1,6-hexanediamine | 65953-56-2 | 99.6% | 342.50 | C16H30N4O2S | 500 MG
SDP
Catalog No. 5000282408
Encompass_Preferred
N-biotinyl-1,6-hexanediamine is a biotinylated biochemical reagent used for labeling and conjugation in life-science research. It provides a primary amine spacer for efficient biotin tagging of molecules, enabling detection, affinity capture, and downstream assay workflows.
  • Biotinylated primary amine spacer for stable conjugation to proteins and small molecules.
  • High purity (99.6%) for consistent experimental performance.
  • Molecular weight 342.50 and formula C16H30N4O2S; CAS 65953-56-2.
  • Suitable for biochemical assays, labeling, and affinity-capture applications.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Available in multiple pack sizes including 500 mg for scale flexibility.
Catalog No. 50-002-82408 Supplier Medchemexpress LLC Supplier No. HYW343597500MG
N-biotinyl-1,6-hexanediamine is a biotinylated biochemical reagent used for labeling and conjugation in life-science research. It provides a primary amine spacer for efficient biotin tagging of molecules, enabling detection, affinity capture, and downstream assay workflows.
  • Biotinylated primary amine spacer for stable conjugation to proteins and small molecules.
  • High purity (99.6%) for consistent experimental performance.
  • Molecular weight 342.50 and formula C16H30N4O2S; CAS 65953-56-2.
  • Suitable for biochemical assays, labeling, and affinity-capture applications.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Available in multiple pack sizes including 500 mg for scale flexibility.