Learn More
Medchemexpress LLC N-biotinyl-1,6-hexanediamine | 65953-56-2 | 99.6% | 342.50 g·mol⁻¹ | C16H30N4O2S | 100 MG
SDP
Catalog No. 5000282562
Encompass_Preferred
N-biotinyl-1,6-hexanediamine is a biotinylated biochemical reagent used for labeling, probe conjugation, and affinity capture in life-science research. It is supplied as a powder of high purity (≈99.6%) with a molecular weight of 342.50 g·mol⁻¹ and has defined storage guidance for solid and solution forms.
  • Biotinylated primary amine for conjugation and labeling
  • High purity suitable for biochemical assays
  • Available in multiple pack sizes for experimental flexibility
  • Stable as a powder at -20°C for long-term storage
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month
  • Accompanied by datasheet, SDS, and COA for quality and safety information
Catalog No. 50-002-82562 Supplier Medchemexpress LLC Supplier No. HYW343597100MG
N-biotinyl-1,6-hexanediamine is a biotinylated biochemical reagent used for labeling, probe conjugation, and affinity capture in life-science research. It is supplied as a powder of high purity (≈99.6%) with a molecular weight of 342.50 g·mol⁻¹ and has defined storage guidance for solid and solution forms.
  • Biotinylated primary amine for conjugation and labeling
  • High purity suitable for biochemical assays
  • Available in multiple pack sizes for experimental flexibility
  • Stable as a powder at -20°C for long-term storage
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month
  • Accompanied by datasheet, SDS, and COA for quality and safety information