Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-biotinyl-1,6-hexanediamine | 65953-56-2 | 99.6% | 342.50 g·mol⁻¹ | C16H30N4O2S | 25 MG
SDP

Catalog No. 5000282407
Encompass_Preferred

N-biotinyl-1,6-hexanediamine is a biotinylated reagent used for labeling and conjugation of biomolecules in biochemical assays and detection workflows. It provides a biotin tag compatible with streptavidin-based capture, detection, and affinity purification.

  • Provides a primary amine-linked biotin tag suitable for conjugation.
  • High purity: 99.6%.
  • Molecular weight 342.50 g·mol⁻¹.
  • CAS number 65953-56-2 for unambiguous identification.
  • Commonly used in labeling, affinity capture, and assay development.

Catalog No. 50-002-82407 Supplier Medchemexpress LLC Supplier No. HYW34359725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-biotinyl-1,6-hexanediamine is a biotinylated reagent used for labeling and conjugation of biomolecules in biochemical assays and detection workflows. It provides a biotin tag compatible with streptavidin-based capture, detection, and affinity purification.

  • Provides a primary amine-linked biotin tag suitable for conjugation.
  • High purity: 99.6%.
  • Molecular weight 342.50 g·mol⁻¹.
  • CAS number 65953-56-2 for unambiguous identification.
  • Commonly used in labeling, affinity capture, and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.