Learn More
Medchemexpress LLC N-biotinyl p-aminophenyl arsenic acid | 212391-23-6 | MFCD29472395 | 98.1% | 427.35 g/mol | C16H22AsN3O4S | 10mg
SDP
Catalog No. 5000270274
Encompass_Preferred
N-Biotinyl p-aminophenyl arsenic acid is a bifunctional reagent that can bind to both streptavidin and dithiols N-Biotinyl p-aminophenyl arsenic acid decreases the Rbungarotoxin-binding sites in reduced Torpedo nicotinic receptors (IC50 is 10-300 nM) and protects the receptor from irreversible alkylation by bromoacetylcholine[1]
Catalog No. 50-002-70274 Supplier Medchemexpress LLC Supplier No. HYW74157310MG
N-Biotinyl p-aminophenyl arsenic acid is a bifunctional reagent that can bind to both streptavidin and dithiols N-Biotinyl p-aminophenyl arsenic acid decreases the Rbungarotoxin-binding sites in reduced Torpedo nicotinic receptors (IC50 is 10-300 nM) and protects the receptor from irreversible alkylation by bromoacetylcholine[1]