Learn More
Medchemexpress LLC N-Bis(2-hydroxypropyl)nitrosamine | 53609-64-6 | 98.0% | 162.19 | 100 MG
SDP
Catalog No. 5000435616
Encompass_Preferred
N-Bis(2-hydroxypropyl)nitrosamine is a tumor model inducing agent and a metabolite of di-n-propylnitrosamine (DPN). It is an agent with carcinogenic activity and can be used to construct pancreatic cancer models. This product is for research use only.
  • Tumor model inducing agent
  • Metabolite of di-n-propylnitrosamine (DPN)
  • Carcinogenic activity
  • Used to construct pancreatic cancer models
Catalog No. 50-004-35616 Supplier Medchemexpress LLC Supplier No. HY112085100MG
N-Bis(2-hydroxypropyl)nitrosamine is a tumor model inducing agent and a metabolite of di-n-propylnitrosamine (DPN). It is an agent with carcinogenic activity and can be used to construct pancreatic cancer models. This product is for research use only.
  • Tumor model inducing agent
  • Metabolite of di-n-propylnitrosamine (DPN)
  • Carcinogenic activity
  • Used to construct pancreatic cancer models