Learn More
Medchemexpress LLC N-Bis(2-hydroxypropyl)nitrosamine | 53609-64-6 | 98.0% | 162.19 | 25 MG
SDP
Catalog No. 5000439511
Encompass_Preferred
N-Bis(2-hydroxypropyl)nitrosamine is a tumor model inducing agent and a metabolite of di-n-propylnitrosamine (DPN). It is used to construct pancreatic cancer models and demonstrates carcinogenic activity. In vivo, it causes tumor development in the lung, liver, and thyroid of rats at a dose of 100 ppm, and in the lung, liver, thyroid, esophagus, kidney, and urinary bladder of rats at a dose of 500 ppm. Bisphenol A (BPA) can enhance the susceptibility to thyroid carcinoma when stimulated by this agent.
  • Tumor model inducing agent.
  • Metabolite of di-n-propylnitrosamine (DPN).
  • Applicable in constructing pancreatic cancer models.
  • Demonstrates carcinogenic activity in vitro.
  • Induces tumor development in various organs in rats.
  • Susceptibility to thyroid carcinoma can be enhanced by Bisphenol A (BPA) stimulation.
Catalog No. 50-004-39511 Supplier Medchemexpress LLC Supplier No. 25MGHY112085
N-Bis(2-hydroxypropyl)nitrosamine is a tumor model inducing agent and a metabolite of di-n-propylnitrosamine (DPN). It is used to construct pancreatic cancer models and demonstrates carcinogenic activity. In vivo, it causes tumor development in the lung, liver, and thyroid of rats at a dose of 100 ppm, and in the lung, liver, thyroid, esophagus, kidney, and urinary bladder of rats at a dose of 500 ppm. Bisphenol A (BPA) can enhance the susceptibility to thyroid carcinoma when stimulated by this agent.
  • Tumor model inducing agent.
  • Metabolite of di-n-propylnitrosamine (DPN).
  • Applicable in constructing pancreatic cancer models.
  • Demonstrates carcinogenic activity in vitro.
  • Induces tumor development in various organs in rats.
  • Susceptibility to thyroid carcinoma can be enhanced by Bisphenol A (BPA) stimulation.