Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-1-(2,4-dinitrophenyl)-L-histidine, 99%, Thermo Scientific™

Catalog No. AAH6342106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6342106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6342106 Supplier Thermo Scientific Chemicals Supplier No. H6342106

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25024-53-7
Molecular Formula C17H19N5O8
Molecular Weight (g/mol) 421.37
MDL Number MFCD00065966
InChI Key KPGVUOQMOHGHEW-LBPRGKRZSA-N
Synonym boc-his dnp-oh, n-tert-butoxycarbonyl-1-2,4-dinitrophenyl-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl, 2s-2-tert-butoxycarbonyl amino-3-1-2,4-dinitrophenyl imidazol-4-yl propanoic acid, pubchem18935, boc-his dnp-oh-ipa, boc-his dnp-oh isopropanol solvate, n-1,1-dimethylethoxy carbonyl-1-2,4-dinitrophenyl-l-histidine, boc-his dnp-oh tlc, n, a-boc-n im-2,4-dinitrophenyl-l-histidine
PubChem CID 90690
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[1-(2,4-dinitrophenyl)-1H-imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=N1)C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Beilstein 771922
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 421.36
Sensitivity Light sensitive
Percent Purity 99%
Physical Form Solid
Chemical Name or Material N-Boc-1-(2,4-dinitrophenyl)-L-histidine
EINECSNumber 246-569-0
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.