Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-1,6-Hexanediamine

Catalog No. 792295G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-229-5G 1 g
79-229-1G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 79-229-5G Supplier Sigma Aldrich Supplier No. 792295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for the introduction of a C6-spacer.

Specifications

CAS 51857-17-1
Density 0.965 g/mL at 20°C
Molecular Formula C11H24N2O2
Linear Formula C11H24N2O2
Refractive Index n20/D 1.462
MDL Number MFCD00671489
Quantity 1 g
Synonym N-Boc-1,6-diaminohexane; tert-Butyl-N-(6-aminohexyl)carbamate
Molecular Weight (g/mol) 216.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.