Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-1-benzyloxymethyl-L-histidine, 97%, Thermo Scientific™

Catalog No. AAH6266306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6266306 5 g
AAH6266303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6266306 Supplier Thermo Scientific Chemicals Supplier No. H6266306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83468-83-1
Molecular Formula C19H25N3O5
Molecular Weight (g/mol) 375.43
MDL Number MFCD00038627
InChI Key IEAOAWZLUGOPJX-UHFFFAOYNA-N
Synonym boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
PubChem CID 11245813
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 375.42
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Boc-1-benzyloxymethyl-L-histidine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.