Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-2-aminoisobutyric acid, 98+%

Catalog No. AAL1948406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948406 5 g
AAL1948403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948406 Supplier Thermo Scientific Chemicals Supplier No. L1948406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 30992-29-1 | C9H17NO4 | 203.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30992-29-1
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00042973
InChI Key MFNXWZGIFWJHMI-UHFFFAOYSA-N
Synonym boc-aib-oh, boc-alpha-methylalanine, 2-boc-amino isobutyric acid, 2-tert-butoxycarbonyl amino-2-methylpropanoic acid, boc-2-aminoisobutyric acid, n-boc-2-aminoisobutyric acid, n-boc-alpha-methylalanine, n-boc-2-amino-2-methylpropanoic acid, 2-tert-butoxy carbonyl amino-2-methylpropanoic acid, 2-tert-butoxycarbonylamino isobutyric acid
PubChem CID 2733845
IUPAC Name 2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(C)(C)C(=O)O

Specifications

Melting Point 118°C to 121°C
Beilstein 1953772
Quantity 5 g
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material N-Boc-2-aminoisobutyric acid
EINECSNumber 250-421-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.