Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-2-aminophenol

Catalog No. 6359605G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-596-05G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-596-05G Supplier Sigma Aldrich Supplier No. 6359605G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Boc-2-aminophenol

Specifications

CAS 186663-74-1
Melting Point 140°C to 144°C (lit.)
Molecular Formula C11H15NO3
Linear Formula HOC6H4NHCO2C(CH3)3
MDL Number MFCD06411300
Quantity 1 g
Synonym 2-Hydroxyphenyl carbamic acid tert-butyl ester; N-tert-Butoxycarbonyl-2-aminophenol
Molecular Weight (g/mol) 209.24
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.