Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-2-chloro-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH6110703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6110703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6110703 Supplier Thermo Scientific Chemicals Supplier No. H6110703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80102-23-4
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.751
MDL Number MFCD00801056
InChI Key TUUQJNHCVFJMPU-LLVKDONJSA-N
Synonym boc-2-chloro-d-phenylalanine, boc-d-2-chlorophe, r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-d-phe 2-cl-oh, boc-d-2-chlorophenylalanine, boc-d-2-chlooro phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-2-chlorophenyl propanoic acid, boc-l-phe 2-cl-oh, d-phenylalanine, 2-chloro-n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxy carbonylamino-3-2-chlorophenyl propanoic acid
PubChem CID 7021188
IUPAC Name (2R)-3-(2-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Cl)C(=O)O

Specifications

Melting Point 94°C to 98°C
Beilstein 8422927
Quantity 1 g
Formula Weight 299.75
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-2-chloro-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.