Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-2-cyclohexyl-L-glycine, 97%, Thermo Scientific™

Catalog No. AAH6246606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6246606 5 g
AAH6246603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6246606 Supplier Thermo Scientific Chemicals Supplier No. H6246606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Boc-2-cyclohexyl-L-glycine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109183-71-3
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD00133638
InChI Key QSUXZIPXYDQFCX-JTQLQIEISA-N
Synonym boc-l-cyclohexylglycine, boc-chg-oh, boc-l-alpha-cyclohexylglycine, n-boc-2-cyclohexyl-l-glycine, s-2-tert-butoxycarbonyl amino-2-cyclohexylacetic acid, boc-cyclohexyl-glycine, 2s-tert-butoxycarbonyl amino cyclohexyl acetic acid, s-tert-butoxycarbonylamino-cyclohexyl-acetic acid, s-tert-butoxycarbonyl amino cyclohexyl acetic acid, 2s-tert-butoxycarbonyl amino cyclohexyl ethanoic acid
PubChem CID 7004938
IUPAC Name (2S)-2-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
SMILES CC(C)(C)OC(=O)NC(C1CCCCC1)C(=O)O

Specifications

Beilstein 5553687
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 257.33
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-2-cyclohexyl-L-glycine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.