Learn More
Medchemexpress LLC N-Boc-2-pyrroleboronic acid | 135884-31-0 | 99.97% | 211.03 | 10 G
SDP
Catalog No. 5000404021
Encompass_Preferred
N-Boc-2-pyrroleboronic acid is an organic synthesis intermediate primarily used in the synthesis of chiral pharmaceutical molecules and biologically active molecules. It is intended for research use only.
  • Boronic acid on the pyrrole ring can participate in Suzuki coupling reactions.
  • Can be oxidized to a hydroxyl group under the action of nitrogen oxides.
  • Can be further transformed into 2-oxo-2,5-dihydro-1-pyrrolecarboxylic acid tert-butyl ester after tautomerization.
Catalog No. 50-004-04021 Supplier Medchemexpress LLC Supplier No. HY3295910G
N-Boc-2-pyrroleboronic acid is an organic synthesis intermediate primarily used in the synthesis of chiral pharmaceutical molecules and biologically active molecules. It is intended for research use only.
  • Boronic acid on the pyrrole ring can participate in Suzuki coupling reactions.
  • Can be oxidized to a hydroxyl group under the action of nitrogen oxides.
  • Can be further transformed into 2-oxo-2,5-dihydro-1-pyrrolecarboxylic acid tert-butyl ester after tautomerization.