Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3,5-difluoro-L-phenylalanine, 95%

Catalog No. AAH62216MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62216MD 250 mg
AAH6221603 1 g
AAH6221606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62216MD Supplier Thermo Scientific Chemicals Supplier No. H62216MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 205445-52-9 | C14H17F2NO4 | 301.29 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205445-52-9
Molecular Formula C14H17F2NO4
Molecular Weight (g/mol) 301.29
MDL Number MFCD00797555
InChI Key CZBNUDVCRKSYDG-NSHDSACASA-N
Synonym boc-phe 3,5-dif-oh, boc-l-3,5-difluorophenylalanine, boc-l-3,5-difluorophe, s-n-boc-3,5-difluorophenylalanine, boc-phe 3,5-f2-oh, boc-3,5-difluoro-l-phenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,5-difluorophenyl propanoic acid, s-2-tert-butoxycarbonyl amino-3-3,5-difluorophenyl propanoic acid, phenylalanine, n-1,1-dimethylethoxy carbonyl-3,5-difluoro, 2s-2-tert-butoxycarbonyl amino-3-3,5-difluorophenyl propionic acid
PubChem CID 2779009
IUPAC Name (2S)-3-(3,5-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC(=C1)F)F)C(=O)O

Specifications

Melting Point 115°C
Quantity 250 mg
Formula Weight 301.29
Percent Purity 95%
Chemical Name or Material N-Boc-3,5-difluoro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.