Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Boc-3-aminothiophene

Catalog No. 7583021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-830-21G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-830-21G Supplier Sigma Aldrich Supplier No. 7583021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Boc-3-aminothiophene

Specifications

CAS 19228-91-2
Melting Point 135°C to 140°C
Molecular Formula C9H13NO2S
Linear Formula C9H13NO2S
MDL Number MFCD01928808
Quantity 1 kg
Synonym 3-(tert-Butoxycarbonylamino)thiophene; 3-Thiophenecarbamic acid tert-butyl ester; N-3-Thienylcarbamic acid 1,1-dimethylethyl ester
Molecular Weight (g/mol) 199.27
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.