Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-benzyloxymethyl-L-histidine, 98%, Thermo Scientific™

Catalog No. AAH6123406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6123406 5 g
AAH6123403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6123406 Supplier Thermo Scientific Chemicals Supplier No. H6123406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79950-65-5
MDL Number MFCD00043133
Synonym Boc-His(Bom)-OH; N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine

Specifications

Color White
Beilstein 4335455
Quantity 5 g
Formula Weight 375.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-3-benzyloxymethyl-L-histidine
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.