Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-cyclohexyl-L-alanine, 98%

Catalog No. AAH5528206
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5528206 5 g
AAH55282MD 250 mg
AAH5528203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5528206 Supplier Thermo Scientific Chemicals Supplier No. H5528206
Only null left
Add to Cart

CAS: 37736-82-6 | C14H25NO4 | 271.357 g/mol

It is an agrochemical, organic and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an agrochemical, organic and pharmaceutical intermediates.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37736-82-6
Molecular Formula C14H25NO4
Molecular Weight (g/mol) 271.357
MDL Number MFCD00211335
InChI Key MSZQAQJBXGTSHP-NSHDSACASA-N
Synonym boc-cha-oh, boc-cha-oh hydrate, s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-beta-cyclohexyl-ala-oh, boc-l-cyclohexylalanine, s-2-tert-butoxycarbonylamino-3-cyclohexyl-propionic acid, n-boc-b-cyclohexylalanine, 2s-2-tert-butoxycarbonyl amino-3-cyclohexylpropanoic acid, boc-cyclohexyl-ala-oh, boc-3-cyclohexyl-l-alanine
PubChem CID 10588189
IUPAC Name (2S)-3-cyclohexyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1CCCCC1)C(=O)O

Specifications

Quantity 5 g
Beilstein 3060603
Formula Weight 271.36
Percent Purity 98%
Chemical Name or Material N-Boc-3-cyclohexyl-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.