Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-BOC-3-fluoro-D-phenylalanine | 114873-11-9 | 99.67% | C14H18FNO4 | 100 G
SDP

Catalog No. 5000433214
Encompass_Preferred

N-BOC-3-Fluoro-D-phenylalanine, also known as N-tert-Butoxycarbonyl-3-fluoro-D-phenylalanine, is a phenylalanine derivative with a molecular weight of 283.30 and chemical formula C14H18FNO4. It appears as a white to off-white solid. Amino acid derivatives are used as ergogenic supplements to influence anabolic hormone secretion, supply fuel during exercise, improve mental performance under stress, and prevent exercise-induced muscle damage.

  • Available with stated purities of 99.48% and 99.67%.
  • Physical state: Solid.
  • Color: White to off-white.
  • Shipping at room temperature within the continental US, with potential variations for other locations.
  • Storage for powder form: -20°C for up to 3 years or at 4°C for up to 2 years.
  • Storage when in solvent: -80°C for up to 6 months or at -20°C for up to 1 month.
  • Exclusively for research purposes; not intended for sale to patients.

Catalog No. 50-004-33214 Supplier Medchemexpress LLC Supplier No. HY79295100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-BOC-3-Fluoro-D-phenylalanine, also known as N-tert-Butoxycarbonyl-3-fluoro-D-phenylalanine, is a phenylalanine derivative with a molecular weight of 283.30 and chemical formula C14H18FNO4. It appears as a white to off-white solid. Amino acid derivatives are used as ergogenic supplements to influence anabolic hormone secretion, supply fuel during exercise, improve mental performance under stress, and prevent exercise-induced muscle damage.

  • Available with stated purities of 99.48% and 99.67%.
  • Physical state: Solid.
  • Color: White to off-white.
  • Shipping at room temperature within the continental US, with potential variations for other locations.
  • Storage for powder form: -20°C for up to 3 years or at 4°C for up to 2 years.
  • Storage when in solvent: -80°C for up to 6 months or at -20°C for up to 1 month.
  • Exclusively for research purposes; not intended for sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.