Learn More
Medchemexpress LLC N-BOC-3-Fluoro-D-phenylalanine | 114873-11-9 | 99.67% | C14H18FNO4 | 5 G
SDP
Catalog No. 5000433063
Encompass_Preferred
N-BOC-3-Fluoro-D-phenylalanine is a phenylalanine derivative intended for research use only. It is not for patient use. Amino acids and their derivatives, including this product, are recognized as beneficial ergogenic dietary substances for their commercial use as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
  • White to off-white solid appearance
  • Molecular weight of 283.30
  • Related to peptides
  • Reagent for peptide synthesis
  • Classified as an amino acid derivative
Catalog No. 50-004-33063 Supplier Medchemexpress LLC Supplier No. HY792955G
N-BOC-3-Fluoro-D-phenylalanine is a phenylalanine derivative intended for research use only. It is not for patient use. Amino acids and their derivatives, including this product, are recognized as beneficial ergogenic dietary substances for their commercial use as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
  • White to off-white solid appearance
  • Molecular weight of 283.30
  • Related to peptides
  • Reagent for peptide synthesis
  • Classified as an amino acid derivative