Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-methyl-L-phenylalanine, 98%

Catalog No. AAH6286006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6286006 5 g
AAH6286003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6286006 Supplier Thermo Scientific Chemicals Supplier No. H6286006

missing translation for 'validMainframeMsg'

CAS: 114873-06-2 | C15H21NO4 | 279.336 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114873-06-2
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD01862944
InChI Key HBBXWMALJNZDOM-LBPRGKRZSA-N
Synonym boc-phe 3-me-oh, boc-3-methyl-l-phenylalanine, boc-l-3-methylphenylalanine, boc-l-3-methylphe, boc-3-methy-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-m-tolyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-methylphenyl propanoic acid, boc-l-3-methyl phenylalanine, boc-phe 3-me-oh tlc, 3-methyl-n-t-butoxycarbonyl-l-phenylalanine
PubChem CID 2761470
IUPAC Name (2S)-3-(3-methylphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC1=CC(=CC=C1)CC(C(=O)O)NC(=O)OC(C)(C)C

Specifications

Beilstein 8989098
Quantity 5 g
Formula Weight 279.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-3-methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.