Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-nitro-L-phenylalanine, 95%

Catalog No. AAH5205803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5205803 1 g
AAH5205806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5205803 Supplier Thermo Scientific Chemicals Supplier No. H5205803
Add to Cart

CAS: 131980-29-5 | C14H18N2O6 | 310.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131980-29-5
Molecular Formula C14H18N2O6
Molecular Weight (g/mol) 310.306
MDL Number MFCD01317033
InChI Key OWTGPXDXLMNQKK-NSHDSACASA-N
Synonym boc-3-nitro-l-phenylalanine, boc-l-3-nitrophenylalanine, boc-phe 3-no2-oh, s-n-boc-3-nitrophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, s-2-tert-butoxycarbonyl amino-3-3-nitrophenyl propanoic acid, boc-l-3-nitrophe, boc-l-3-no2-phe-oh, 2-tert-butoxycarbonylamino-3-3-nitro-phenyl-propionic acid
PubChem CID 2761816
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Quantity 1 g
Formula Weight 310.31
Percent Purity 95%
Chemical Name or Material N-Boc-3-nitro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.