Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-pyrroline, 97%, Thermo Scientific™

Catalog No. AAL2040603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2040603 1 g
AAL2040606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2040603 Supplier Thermo Scientific Chemicals Supplier No. L2040603

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in a synthesis of ß-aryl-GABA analogues by Heck arylation with arenediazonium salts, as well as synthesize aryl pyrrolizidines. N-Boc-pyrroline has also been utilized in the synthesis of various antibacterial compounds. It has also been used in the preparation of regioisomeric 3-hydroxyisoxazolinyl prolines which are medicinally active.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73286-70-1
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.224
MDL Number MFCD01863512
InChI Key YEBDZDMYLQHGGZ-UHFFFAOYSA-N
Synonym tert-butyl 2,5-dihydro-1h-pyrrole-1-carboxylate, n-boc-2,5-dihydropyrrole, n-boc-3-pyrroline, 1-boc-2,5-dihydro-1h-pyrrole, tert-butyl 2h-pyrrole-1 5h-carboxylate, n-boc-2,5-dihydro-1h-pyrrole, 2,5-dihydro-pyrrole-1-carboxylic acid tert-butyl ester, 3-pyrroline, n-boc protected, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, 1,1-dimethylethyl ester, tert-butyl-2,5-dihydro-1h-pyrrole-1-carboxylate
PubChem CID 5148195
IUPAC Name tert-butyl 2,5-dihydropyrrole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC=CC1

Specifications

Boiling Point 208°C
Refractive Index 1.458
Solubility Information Slightly soluble in water.
Density 0.981
Quantity 1 g
Flash Point 81°C (178°F)
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material N-Boc-3-pyrroline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.