Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-3-trifluoromethyl-L-phenylalanine, 98%, Thermo Scientific™

Catalog No. AAH6263703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6263703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6263703 Supplier Thermo Scientific Chemicals Supplier No. H6263703

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142995-31-1
Molecular Formula C15H18F3NO4
Molecular Weight (g/mol) 333.307
MDL Number MFCD00672524
InChI Key SHMWDGGGZHFFRC-NSHDSACASA-N
Synonym boc-phe 3-cf3-oh, boc-l-3-trifluoromethylphenylalanine, boc-3-trifluoromethyl-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-3-trifluoromethyl phenyl propanoic acid, boc-l-3-trifluoromethylphe, boc-d-3-trifluoromethylphenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3-trifluoromethyl phenyl propanoic acid, s-2-tert-butoxycarbonylamino-3-3-trifluoromethyl-phenyl-propionic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-3-3-trifluoromethyl phenyl propanoic acid
PubChem CID 7020838
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(trifluoromethyl)phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 135°C to 138°C
Beilstein 7048145
Quantity 1 g
Formula Weight 333.3
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Boc-3-trifluoromethyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.