Learn More
Medchemexpress LLC N-Boc-4-bromopiperidine | 180695-79-8 | 264.16 | 25 G
SDP
Catalog No. 5000449944
Encompass_Preferred
N-Boc-4-bromopiperidine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Appearance: White to light yellow
  • Physical state: Solid below 42°C, liquid above 42°C
Catalog No. 50-004-49944 Supplier Medchemexpress LLC Supplier No. 25GHYW001478
N-Boc-4-bromopiperidine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Appearance: White to light yellow
  • Physical state: Solid below 42°C, liquid above 42°C