Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-4-fluoro-L-phenylalanine, 95%

Catalog No. AAH6137006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6137006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6137006 Supplier Thermo Scientific Chemicals Supplier No. H6137006

CAS: 41153-30-4 | C14H18FNO4 | 283.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41153-30-4
Molecular Formula C14H18FNO4
Molecular Weight (g/mol) 283.299
MDL Number MFCD00079672
InChI Key RCXSXRAUMLKRRL-NSHDSACASA-N
Synonym boc-l-4-fluorophenylalanine, boc-phe 4-f-oh, boc-l-4-fluorophe, boc-4-fluoro-l-phenylalanine, boc-4-fluoro-l-phe-oh, n-tert-butoxycarbonyl-4-fluoro-l-phenylalanine, s-n-boc-4-fluorophenylalanine, boc-l-phe 4-f-oh, boc-p-fluoro-l-phenylalanine, boc-p-fluoro-phe-oh
PubChem CID 7020910
IUPAC Name (2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O

Specifications

Melting Point 83°C to 86°C
Color White
Beilstein 4323475
Quantity 5 g
Formula Weight 283.3
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-4-fluoro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.