Learn More
Medchemexpress LLC HY-43721 10mg Medchemexpress, N-Boc-4-pentyne-1-amine CAS:151978-50-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY4372110MG
Encompass_Preferred
Medchemexpress, HY-43721 10mg N-Boc-4-pentyne-1-amine CAS:151978-50-6 N-Boc-4-pentyne-1-amine is a PROTAC linker, which refers to the alkyl chain composition. N-Boc-4-pentyne-1-amine can be used in the synthesis of the PROTAC MG-277 (HY-130122). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0059