Learn More
Medchemexpress LLC N-Boc-4-piperidinemethanol | 123855-51-6 | 99.91% | 215.29 | 25 G
SDP
Catalog No. 5000402868
Encompass_Preferred
N-Boc-4-piperidinemethanol is a chemical compound primarily recognized as an acetylcholinesterase (AChE) inhibitor. This compound is of interest in research, particularly in studies related to neurological diseases such as Alzheimer's Disease. Its molecular structure includes a piperidine ring with a protected hydroxylmethyl group, making it a versatile building block in organic synthesis and drug discovery.
  • Functions as an acetylcholinesterase inhibitor.
  • Utilized in research for neurological diseases.
  • Features a piperidine ring structure.
  • Suitable for organic synthesis and drug discovery applications.
Catalog No. 50-004-02868 Supplier Medchemexpress LLC Supplier No. 25GHY21629
N-Boc-4-piperidinemethanol is a chemical compound primarily recognized as an acetylcholinesterase (AChE) inhibitor. This compound is of interest in research, particularly in studies related to neurological diseases such as Alzheimer's Disease. Its molecular structure includes a piperidine ring with a protected hydroxylmethyl group, making it a versatile building block in organic synthesis and drug discovery.
  • Functions as an acetylcholinesterase inhibitor.
  • Utilized in research for neurological diseases.
  • Features a piperidine ring structure.
  • Suitable for organic synthesis and drug discovery applications.