Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-4-Piperidinemethanol, 97%

Catalog No. p-3771119
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397720010 1 g Glass bottle
AC397720050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397720010 Supplier Thermo Scientific Chemicals Supplier No. 397720010
Only null left
Add to Cart

CAS: 123855-51-6 | C11H21NO3 | 215.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123855-51-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD02094488
InChI Key CTEDVGRUGMPBHE-UHFFFAOYSA-N
Synonym n-boc-4-piperidinemethanol, tert-butyl 4-hydroxymethyl piperidine-1-carboxylate, 1-boc-4-piperidinemethanol, n-boc-4-hydroxymethylpiperidine, tert-butyl 4-hydroxymethyl-1-piperidinecarboxylate, 1-tert-butoxycarbonyl-4-hydroxymethyl piperidine, 1-n-boc-4-hydroxymethyl-piperidine, n-boc-4-hydroxylmethylene-piperidine, 1-tert-butoxycarbonyl-4-piperidinemethanol, n-tert-butoxycarbonyl-4-piperidinemethanol
PubChem CID 2764081
IUPAC Name tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CO)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 78°C to 82°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-BOC-4-Piperidinemethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.