Learn More
eMolecules 400727-63-1 | N-Boc-5-aminoisoindoline | MFCD08236255 | 1g
Supplier: eMolecules 599169687
ChemScene | (4-Methanesulfonylphenyl)acetic acid | 10g | 536831957 | CS-B0763 | 90536-66-6 | MFCD00216495 | 214.240 | C9H10O4S
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool<|a>
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.