Learn More
Medchemexpress LLC N-Boc-7-aminoheptanoic acid | 60142-89-4 | MFCD00063356 | ≥98.0% | 245.32 | 50 G
SDP
Catalog No. 5000452293
Encompass_Preferred
N-Boc-7-aminoheptanoic acid is a synthetic amino acid derivative featuring a Boc-protected amine group and a terminal carboxylic acid. This compound is widely utilized in organic synthesis, particularly as a PROTAC linker, facilitating the construction of proteolysis-targeting chimeras (PROTACs) and other conjugation applications. Its structure allows for selective modification and plays a crucial role in drug discovery and chemical biology research.
  • Acts as a versatile PROTAC linker
  • Features a Boc-protected amine group for selective reactions
  • Essential for the synthesis of advanced chemical compounds
  • Purity of ≥98.0% ensures reliable experimental results
Catalog No. 50-004-52293 Supplier Medchemexpress LLC Supplier No. HYW01200150G
N-Boc-7-aminoheptanoic acid is a synthetic amino acid derivative featuring a Boc-protected amine group and a terminal carboxylic acid. This compound is widely utilized in organic synthesis, particularly as a PROTAC linker, facilitating the construction of proteolysis-targeting chimeras (PROTACs) and other conjugation applications. Its structure allows for selective modification and plays a crucial role in drug discovery and chemical biology research.
  • Acts as a versatile PROTAC linker
  • Features a Boc-protected amine group for selective reactions
  • Essential for the synthesis of advanced chemical compounds
  • Purity of ≥98.0% ensures reliable experimental results