Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-8-amino-octanoic acid | 30100-16-4 | MFCD00270350 | >=97.0% | 259.34 g/mol | C13H25NO4 | 10 G
SDP

Catalog No. 5000209174
Encompass_Preferred

N-Boc-8-amino-octanoic acid is a Boc-protected amino acid used as a linker building block in PROTAC and peptide synthesis. It contains a terminal carboxylic acid and a tert-butyloxycarbonyl (Boc)-protected amine, suitable for amide coupling and mild acidic deprotection.

  • Molecular formula: C13H25NO4
  • Molecular weight: 259.34 g/mol
  • Cas number: 30100-16-4
  • Purity: ≥97.0%
  • Appearance: white to off-white solid
  • Storage: store at room temperature; in solvent store at -80°C (2 years) or -20°C (1 year)
  • Typical applications: protac linker and peptide or small-molecule synthesis

Catalog No. 50-002-09174 Supplier Medchemexpress LLC Supplier No. HYW04185610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Boc-8-amino-octanoic acid is a Boc-protected amino acid used as a linker building block in PROTAC and peptide synthesis. It contains a terminal carboxylic acid and a tert-butyloxycarbonyl (Boc)-protected amine, suitable for amide coupling and mild acidic deprotection.

  • Molecular formula: C13H25NO4
  • Molecular weight: 259.34 g/mol
  • Cas number: 30100-16-4
  • Purity: ≥97.0%
  • Appearance: white to off-white solid
  • Storage: store at room temperature; in solvent store at -80°C (2 years) or -20°C (1 year)
  • Typical applications: protac linker and peptide or small-molecule synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.