Learn More
Medchemexpress LLC N-Boc-8-amino-octanoic acid | 30100-16-4 | MFCD00270350 | >97.0% | 259.34 g/mol | C13H25NO4 | 5 G
SDP
Catalog No. 5000209558
Encompass_Preferred
N-Boc-8-amino-octanoic acid is a Boc-protected amino acid used as a linker and building block in small-molecule synthesis, including proteolysis-targeting chimera (PROTAC) construction. It contains a Boc-protected terminal amine and a terminal carboxylic acid that can be activated for amide formation.
  • Boc-protected terminal amine for temporary protection
  • Terminal carboxylic acid suitable for activation and amide coupling
  • High purity (>97.0%) suitable for synthetic applications
  • Available in small pack sizes, including 5 G
  • Stable as a solid at room temperature for extended storage
Catalog No. 50-002-09558 Supplier Medchemexpress LLC Supplier No. HYW0418565G
N-Boc-8-amino-octanoic acid is a Boc-protected amino acid used as a linker and building block in small-molecule synthesis, including proteolysis-targeting chimera (PROTAC) construction. It contains a Boc-protected terminal amine and a terminal carboxylic acid that can be activated for amide formation.
  • Boc-protected terminal amine for temporary protection
  • Terminal carboxylic acid suitable for activation and amide coupling
  • High purity (>97.0%) suitable for synthetic applications
  • Available in small pack sizes, including 5 G
  • Stable as a solid at room temperature for extended storage